Learn More
Electron Microscopy Sciences Magnifier 10x Lighted
SDP
Supplier:  Electron Microscopy Sciences 68122
Encompass_Preferred
simple and accurate comparator, high quality measuring scale, detachable flashlight, magnification: 10X, size: 47mm(dia) x 45mm(H) x 185mm(L), scale diameter: 32mm;.
Catalog No. 50-269-38