Learn More
STEMCELL Technologies EasySep™ Red Blood Cell Lysis Buffer, Size: 10 mL
SDP
Catalog No. NC9073463
Encompass
EasySep™ Red Blood Cell Lysis Buffer is for use as directed with EasySep™ or RoboSep™ Whole Blood kits for the lysis of red blood cells prior to separation from whole blood.
Catalog No. NC9073463 Supplier STEMCELL Technologies Supplier No. 20110
EasySep™ Red Blood Cell Lysis Buffer is for use as directed with EasySep™ or RoboSep™ Whole Blood kits for the lysis of red blood cells prior to separation from whole blood.