Learn More
Teknova 10X MOPS Buffer. 100mL, Sterile.
SDP
Catalog No. NC9486905
Encompass
MOPS (3-(N-morpholino)propanesulfonic acid) is a Good’s buffer and has a pKa of 7.01, making it an ideal buffer for many biological systems at near-neutral pH. MOPS can be used in cell culture media and biopharma buffer formulations as well as in the purification of antibodies, peptides, proteins and blood components. It is nontoxic to cell culture lines.
Catalog No. NC9486905 Supplier Teknova Supplier No. M2101
MOPS (3-(N-morpholino)propanesulfonic acid) is a Good’s buffer and has a pKa of 7.01, making it an ideal buffer for many biological systems at near-neutral pH. MOPS can be used in cell culture media and biopharma buffer formulations as well as in the purification of antibodies, peptides, proteins and blood components. It is nontoxic to cell culture lines.