Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ 10X PBS

Catalog No. 48705006
Change view
Click to view available options
Quantity:
3 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-705-006 3 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 48-705-006 Supplier R&D Systems™ Supplier No. 48705006
Add to Cart
Add to Cart
This item is not returnable. View return policy

The 10X PBS (phosphate buffered saline) buffer is a high quality concentrated solution manufactured without magnesium or calcium, and is suitable for cell resuspension and washing in the in situ apoptosis detection (TUNEL) procedure, CometAssay™ (Cat No. 4250-50-K) applications, and any other experiment requiring ultrapure PBS buffer.

Formulation:

  • 75 mM disodium hydrogen phosphate
  • 25 mM sodium dihydrogen phosphate
  • 1.45 M sodium chloride
  • pH 7.4
  • Sterility tested

Specifications

Quantity 3 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.