Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PBS - Phosphate-Buffered Saline (10X) pH 7.4, RNase-free

Catalog No. am9625
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM9625 1 L
AM9624 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AM9625 Supplier Invitrogen™ Supplier No. AM9625
Only null left
Add to Cart
Edge
Add to Cart

PBS (phosphate buffered saline) is a pH-adjusted blend of ultrapure-grade phosphate buffers and saline solutions.

PBS (phosphate buffered saline) is a pH-adjusted blend of ultrapure-grade phosphate buffers and saline solutions which, when diluted to a 1X working concentration, contains 137 mM NaCl, 2.7 mM KCl, 8 mM Na2HPO4, and 2 mM KH2PO4. Each 10X PBS solution is ready to use upon dilution to the desired concentration. This molecular biology-grade PBS is certified RNase-free; it is rigorously tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity.

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phosphate Buffered Saline (PBS)
Concentration 10 X
pH 7.4
Physical Form Liquid
Product Line Ambion
Quantity 1 L
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.