Learn More
Rockland Immunochemicals 10X PBS pH 7.2 (0.2 M Potassium Phosphate 1.5 M Sodium Chloride) (4 x 1 liter) - MB-008-4000
SDP
Catalog No. 501058971
Encompass_Preferred
Phosphate buffered saline is suitable for multiple applications including biological diluent buffer for antibodies or other biologics. Also may be used as a wash buffer for immunological assays including western blot, immunohistochemistry, immunofluorescence microscopy, and ELISA. Other applications may require detergents or other additional components.
Catalog No. 50-105-8971 Supplier Rockland Immunochemicals Supplier No. MB0084000
Phosphate buffered saline is suitable for multiple applications including biological diluent buffer for antibodies or other biologics. Also may be used as a wash buffer for immunological assays including western blot, immunohistochemistry, immunofluorescence microscopy, and ELISA. Other applications may require detergents or other additional components.