Learn More
Rockland Immunochemicals 10X PBS pH 7.2 with Azide (0.2 M Potassium Phosphate 1.5 M Sodium Chloride 0.1% (w/v) Sodium Azide) - MB-011
SDP
Catalog No. 501058976
Encompass_Preferred
Phosphate Buffered Saline (PBS) is a regularly used buffer in biological research. The phosphate ions help maintain a constant pH around neutral. The osmolarity of solution typically matches that of the human body (isotonic solution). PBS can act as a diluent for many experimental preparations and is non-toxic to cells. This preparation contains 0.1% sodium azide as a preservative and is 10X concentrated to be diluted 10-fold to the working concentration.
Catalog No. 50-105-8976 Supplier Rockland Immunochemicals Supplier No. MB011
Phosphate Buffered Saline (PBS) is a regularly used buffer in biological research. The phosphate ions help maintain a constant pH around neutral. The osmolarity of solution typically matches that of the human body (isotonic solution). PBS can act as a diluent for many experimental preparations and is non-toxic to cells. This preparation contains 0.1% sodium azide as a preservative and is 10X concentrated to be diluted 10-fold to the working concentration.