Learn More
Teknova 10X PBS Solution, pH 7.4 at 1X. 4L, Non-Sterile.
SDP
Catalog No. 502388145
Encompass
Phosphate-buffered saline (PBS) is a salt solution that is commonly used in physiological and biochemical experiments. It can be used to maintain pH neutrality or to buffer against changes in pH in order to provide optimal conditions for cellular growth and function. The buffering range is between pH 6.8 and 7.4.
Catalog No. 50-238-8145 Supplier Teknova Supplier No. P0496
Add to Cart
Add to Cart
Phosphate-buffered saline (PBS) is a salt solution that is commonly used in physiological and biochemical experiments. It can be used to maintain pH neutrality or to buffer against changes in pH in order to provide optimal conditions for cellular growth and function. The buffering range is between pH 6.8 and 7.4.