Learn More
Seracare Life Sciences Inc 10X PBS W/ TWEEN 20(PBST)200ML
Catalog No. 5067478
Encompass_Preferred
10X Phosphate-buffered Saline with Tween 20 (PBST) is ideal for use as a wash buffer in general immunoassay procedures. Tween 20, a non-ionic detergent, reduces non-specific binding and protein-protein interaction during the wash step in immunoassay procedures. pH=7.4. Conductivity controlled.
This item is not returnable. View return policy
Catalog No. 50-674-78 Supplier Seracare Life Sciences Inc Supplier No. 54600026
10X Phosphate-buffered Saline with Tween 20 (PBST) is ideal for use as a wash buffer in general immunoassay procedures. Tween 20, a non-ionic detergent, reduces non-specific binding and protein-protein interaction during the wash step in immunoassay procedures. pH=7.4. Conductivity controlled.