Learn More
Edmund Optics 10X PEAK LOUPE, W/LINEAR SCALE
Catalog No. NC0701359
Encompass
10X Peak Measuring Loupe with linear scale. Includes removable glass reticle scale that should be placed in direct contact with object. Features smooth helicoid twist focus and achromatic lens system for color correct flat field viewing.
Catalog No. NC0701359 Supplier Edmund Optics Supplier No. 88614
10X Peak Measuring Loupe with linear scale. Includes removable glass reticle scale that should be placed in direct contact with object. Features smooth helicoid twist focus and achromatic lens system for color correct flat field viewing.