Learn More
Seracare Life Sciences Inc 10X Phosphate Buffered Saline (PBS), 5 Liter
Catalog No. 5067477
Encompass_Preferred
10X Phosphate-buffered Saline (PBS) is ideal for use in sample preparation and as a wash buffer in general immunoassay applications. pH=7.4. Conductivity controlled. 5L
This item is not returnable. View return policy
Catalog No. 50-674-77 Supplier Seracare Life Sciences Inc Supplier No. 54600023
10X Phosphate-buffered Saline (PBS) is ideal for use in sample preparation and as a wash buffer in general immunoassay applications. pH=7.4. Conductivity controlled. 5L