Learn More
Seracare Life Sciences Inc 10X PHOSPHATE BUFFERED SALINE
Catalog No. 5067476
Encompass_Preferred
10X Phosphate-buffered Saline (PBS) is ideal for use in sample preparation and as a wash buffer in general immunoassay applications. pH=7.4. Conductivity controlled.
This item is not returnable. View return policy
Catalog No. 50-674-76 Supplier Seracare Life Sciences Inc Supplier No. 54600022
10X Phosphate-buffered Saline (PBS) is ideal for use in sample preparation and as a wash buffer in general immunoassay applications. pH=7.4. Conductivity controlled.