Learn More
Leinco Technologies Inc 10X Preservative Free ClearSort™ Sheath Fluid | 4 x 1L
SDP
Catalog No. NC0400362
Encompass_Preferred
ClearSort™ 10X Preservative-free Sheath Fluid is specifically formulated to be the highest quality product available for cell sorting instruments. This phosphate buffered saline solution is shipped sterile in case options of 4X1L or bulk 4X4L cases and is designed for use in cell sorters and applications requiring a sterile PBS solution.
Catalog No. NC0400362 Supplier Leinco Technologies Inc Supplier No. S632 4X1L
ClearSort™ 10X Preservative-free Sheath Fluid is specifically formulated to be the highest quality product available for cell sorting instruments. This phosphate buffered saline solution is shipped sterile in case options of 4X1L or bulk 4X4L cases and is designed for use in cell sorters and applications requiring a sterile PBS solution.