Learn More
BIOGEMS INTERNATIONAL INC 10X RBC Lysis Buffer 100 ml
SDP
Catalog No. 501530222
Encompass
Red Blood Cell Lysing Buffer, containing ammonium chloride, is formulated to lyse RBC from human and murine samples while leaving the lymphocytes intact. This buffer, supplied at 10X dilution, should be diluted to 1X to produce the desired effect.
Catalog No. 50-153-0222 Supplier BIOGEMS INTERNATIONAL INC Supplier No. 6401000100
Red Blood Cell Lysing Buffer, containing ammonium chloride, is formulated to lyse RBC from human and murine samples while leaving the lymphocytes intact. This buffer, supplied at 10X dilution, should be diluted to 1X to produce the desired effect.