Learn More
Rockland Immunochemicals 10X SDS-PAGE Running Gel Buffer (0.25 M Tris, 1.92 M Glycine, 1.0% SDS pH 8.3) - MB-017
SDP
Catalog No. 501058981
Encompass_Preferred
SDS-PAGE, sodium dodecyl sulfate polyacrylamide gel electrophoresis, describes a technique widely used in biochemistry to separate proteins according to their electrophoretic mobility (a function of the length of a polypeptide chain and its charge) and no other physical feature. SDS is an anionic detergent applied to protein sample to linearize proteins and to impart a negative charge to linearized proteins. 10X SDS-PAGE Running Buffer is suitable for laboratory involved in protein biochemistry. Visit our newly expanded web site at www.rockland-inc.com for methods using this and other buffers.
Catalog No. 50-105-8981 Supplier Rockland Immunochemicals Supplier No. MB017
SDS-PAGE, sodium dodecyl sulfate polyacrylamide gel electrophoresis, describes a technique widely used in biochemistry to separate proteins according to their electrophoretic mobility (a function of the length of a polypeptide chain and its charge) and no other physical feature. SDS is an anionic detergent applied to protein sample to linearize proteins and to impart a negative charge to linearized proteins. 10X SDS-PAGE Running Buffer is suitable for laboratory involved in protein biochemistry. Visit our newly expanded web site at www.rockland-inc.com for methods using this and other buffers.