Learn More
Teknova 10X SPG (Sucrose Phosphate Glutamate) Solution, 1000mL. Non-Sterile. 2 pack.
SDP
Supplier:  Teknova S00622PK
Encompass
38mM Potassium Phosphate Monobasic,72mM Potassium Phosphate Dibasic,2.18M Sucrose,49mM L-Glutamic Acid Free Acid
Catalog No. 50-238-7290