Learn More
R&D Systems™ Cultrex Spheroid Formation Extracellular Matrix, 10X

Description
- Source: Murine Engelbreth-Holm-Swarm (EHS) tumor.
- Storage Buffer: Dulbecco’s Modified Eagle’s medium (DMEM) containing 10 μg/mL gentamicin sulfate
- Storage/Stability: Product is stable for a minimum of 3 months from date of shipment. Store at –80°C; keep frozen. Dispense into working aliquots if needed; repeated freeze-thaws will destroy product integrity
Specifications
Specifications
| Content And Storage | Store the unopened product at -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Quantity | 600 μL |
| Species | Mouse |
| Source | Murine Engelbreth-Holm-Swarm (EHS) tumor |
| Target | Basement Membrane Extracts |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.