Learn More
ProteinSimple 10x System Control Primary Antibody (Rabbit), 250 uL
Supplier:  ProteinSimple 042196
Encompass
Compatible with Jess, Abby, Wes, Peggy Sue and Sally Sue systems. To be used with rabbit primary antibody to Antibody Diluent. Detects system controls. Match with Standard pack 1, 2, 3 or 4 (250 ul/vial).
Catalog No. NC0697872