Learn More
ProteinSimple 10x System Control Primary Antibody (Mouse), 250 uL
Supplier:  ProteinSimple 042191
Encompass
Compatible with Jess, Abby, Wes, Peggy Sue and Sally Sue systems. The 10X System Control Primary Antibody-Mouse detects system controls is to be used with Antibody Diluent. Please match accordingly with Standard packs 1, 2, 3 or 4 (250 ul/vial).
Catalog No. NC0697892