Learn More
IBI SCIENTIFIC 3P 10X TBE Buffer, 1 liter: Superior Buffering Capacity for Electrophoresis Applications
Catalog No. NC4031743
Encompass_Preferred
TBE (Tris / Boric Acid / EDTA) buffer is a high-performance solution designed to offer superior buffering capacity compared to TAE. With its enhanced ability to maintain pH stability over extended electrophoresis runs, TBE is the preferred choice for DNA sequencing, gel electrophoresis, and other applications requiring high-resolution separation. This buffer eliminates the need for recirculation in most electrophoresis protocols, making it more efficient and reliable for laboratories conducting extended gel runs.
Catalog No. NC4031743 Supplier IBI SCIENTIFIC 3P Supplier No. IB70150
TBE (Tris / Boric Acid / EDTA) buffer is a high-performance solution designed to offer superior buffering capacity compared to TAE. With its enhanced ability to maintain pH stability over extended electrophoresis runs, TBE is the preferred choice for DNA sequencing, gel electrophoresis, and other applications requiring high-resolution separation. This buffer eliminates the need for recirculation in most electrophoresis protocols, making it more efficient and reliable for laboratories conducting extended gel runs.