Learn More
Bio Rad Laboratories 10X TBE BUFFER 1 LTR
Catalog No. NC9372857
Encompass
Reagents; Molecular Biology; 10X Tris/Boric Acid/EDTA; Bio-Rad; Premixed Electrophoresis Buffers; 1L; Made with electrophoresis purity reagents and are quality controlled to ensure reproducible results
Catalog No. NC9372857 Supplier Bio Rad Laboratories Supplier No. 1610733
Reagents; Molecular Biology; 10X Tris/Boric Acid/EDTA; Bio-Rad; Premixed Electrophoresis Buffers; 1L; Made with electrophoresis purity reagents and are quality controlled to ensure reproducible results