Learn More
Teknova 10X TBE DNA Sequencing Grade Solution. 1000mL, Non-Sterile 2 pack.
SDP
Catalog No. 502387251
Encompass
890mM Tris,890mM Boric Acid, 20mM Ethylenediaminetetraacetic acid (EDTA)
Catalog No. 50-238-7251 Supplier Teknova Supplier No. T02052PK
890mM Tris,890mM Boric Acid, 20mM Ethylenediaminetetraacetic acid (EDTA)