Learn More
Growcells 10X TBS 10L EA
Catalog No. 501034864
Encompass_Preferred
Sterile Tris-buffered Saline (TBS) prepared with 18.2 megOhm water, 0.2 micron sterile filtered and steam sterilized in final packaging. Recipe/L: NaCl 80g, KCl 2g, Tris Base 30g. pH 7.4
Catalog No. 50-103-4864 Supplier Growcells Supplier No. MRGF6690010L
Sterile Tris-buffered Saline (TBS) prepared with 18.2 megOhm water, 0.2 micron sterile filtered and steam sterilized in final packaging. Recipe/L: NaCl 80g, KCl 2g, Tris Base 30g. pH 7.4