Learn More
Teknova 10X TBS, pH 7.4. 1000mL, Sterile (247mM Tris, 27mM KCl, 1.37M NaCl) 2 pack.
SDP
Catalog No. 502387596
Encompass
247mM Tris-HCl,27mM Potassium Chloride,1.37M Sodium Chloride
Catalog No. 50-238-7596 Supplier Teknova Supplier No. T95302PK
247mM Tris-HCl,27mM Potassium Chloride,1.37M Sodium Chloride