Learn More
Teknova 10X TBS with 0.5% Tween-20. (247mM Tris, 27mM KCl, 1.37M NaCl) 1000mL, Sterile. 8 pack.
SDP
Supplier:  Teknova T0310PK
Encompass
10X TBS with 0.5% Tween-20. (247mM Tris, 27mM KCl, 1.37M NaCl) 1000mL, Sterile. 8 pack.
Catalog No. 50-227-4066