Learn More
Teknova 10X TBS with 0.5% Tween-20. (247mM Tris, 27mM KCl, 1.37M NaCl) 1000mL, Sterile.
SDP
Catalog No. 502388193
Encompass
Tris (tris(hydroxymethyl)aminomethane) is used as a component ofbuffer solutions, especially for solutions ofnucleic acids. The useful buffer range for tris (pH 7–9) is similar to the pH of most living organisms. This, and its low cost, make tris one of the most common buffers in the biology/biochemistry laboratory.
Catalog No. 50-238-8193 Supplier Teknova Supplier No. T0310
Add to Cart
Add to Cart
Tris (tris(hydroxymethyl)aminomethane) is used as a component ofbuffer solutions, especially for solutions ofnucleic acids. The useful buffer range for tris (pH 7–9) is similar to the pH of most living organisms. This, and its low cost, make tris one of the most common buffers in the biology/biochemistry laboratory.