Learn More
Boston Bioproducts Inc Tris Buffered Saline-Tween, TBST (10X, with 0.5% Tween-20, pH 7.4) - 1L
Catalog No. NC9195866
Encompass_Preferred
10X Tris Buffered Saline - Tween (TBST) is a buffer-surfactant solution supporting a stable pH range between 7.4 and 8.0. When combined in a variable ratio, TBST is an effective wash buffer in Western blotting, in situ hybridization procedures, and immunohistochemistry staining, as it minimizes non specific binding of the materials without disrupting antigen-antibody binding. TBST is also used as an antibody diluent in immunoassays, and supports an array of molecular biology techniques.
Catalog No. NC9195866 Supplier Boston Bioproducts Inc Supplier No. IBB1811L
10X Tris Buffered Saline - Tween (TBST) is a buffer-surfactant solution supporting a stable pH range between 7.4 and 8.0. When combined in a variable ratio, TBST is an effective wash buffer in Western blotting, in situ hybridization procedures, and immunohistochemistry staining, as it minimizes non specific binding of the materials without disrupting antigen-antibody binding. TBST is also used as an antibody diluent in immunoassays, and supports an array of molecular biology techniques.