Learn More
Teknova 10X TBST, pH 7.6, 4L, Non-Sterile.
SDP
Catalog No. 502388067
Encompass
200mM Tris-HCl,1.38M Sodium Chloride,1.0% Tween-20
Catalog No. 50-238-8067 Supplier Teknova Supplier No. T9511
200mM Tris-HCl,1.38M Sodium Chloride,1.0% Tween-20