Learn More
Teknova 10X TE Buffer Solution. 250mL, DNase/RNase, PCR Grade, Sterile. 4 pack.
SDP
Catalog No. 502387773
Encompass
TE buffer is a salt solution that is commonly used to resuspend (or dilute) DNA and RNA. Composed of Tris base and EDTA, these molecules work together to maintain pH and prevent nucleic acids from degradation.
Catalog No. 50-238-7773 Supplier Teknova Supplier No. T34574PK
TE buffer is a salt solution that is commonly used to resuspend (or dilute) DNA and RNA. Composed of Tris base and EDTA, these molecules work together to maintain pH and prevent nucleic acids from degradation.