Learn More
Teknova 10X TE Buffer Solution. DNase/RNase Tested, PCR Grade. 100mL, Sterile.
SDP
Catalog No. 502387689
Encompass
TE buffer is a salt solution that is commonly used to resuspend (or dilute) DNA and RNA. Composed of Tris base and EDTA, these molecules work together to maintain pH and prevent nucleic acids from degradation.
Catalog No. 50-238-7689 Supplier Teknova Supplier No. T3456
Add to Cart
Add to Cart
TE buffer is a salt solution that is commonly used to resuspend (or dilute) DNA and RNA. Composed of Tris base and EDTA, these molecules work together to maintain pH and prevent nucleic acids from degradation.