Learn More
Rockland Immunochemicals 10X TE pH 8.0 (100 mM Tris HCl 10 mM EDTA) DEPC TREATED - MB-007
SDP
Catalog No. 501058969
Encompass_Preferred
TE buffer is a commonly used buffer solution in molecular biology, especially in procedures involving DNA, cDNA or RNA. "TE" is derived from its components: Tris, a common pH buffer, and EDTA, a molecule that chelates cations like Mg2+. The purpose of TE buffer is to solubilize DNA or RNA, while protecting it from degradation.
Catalog No. 50-105-8969 Supplier Rockland Immunochemicals Supplier No. MB007
TE buffer is a commonly used buffer solution in molecular biology, especially in procedures involving DNA, cDNA or RNA. "TE" is derived from its components: Tris, a common pH buffer, and EDTA, a molecule that chelates cations like Mg2+. The purpose of TE buffer is to solubilize DNA or RNA, while protecting it from degradation.