Learn More
Boston Bioproducts Inc Tris Buffered Saline, TBS (10X, for Western Blot Washing) - 1L
Catalog No. NC2145799
Encompass
The Tris Buffered Saline (TBS) is an isotonic non-toxic buffer solution used commonly in biochemical applications. Tris maintains a stable pH range typically between 7.4 to 8.0 while NaCl regulates osmotic balance via its elected concentration. Tris maintains a stable pH range typically between 7.4 to 8.0 while NaCl regulates osmotic balance via its elected concentration. This TBS Solution is formulated at a 10X concentration, and is useful for washing in Western Blotting.
This item is not returnable. View return policy
Catalog No. NC2145799 Supplier Boston Bioproducts Inc Supplier No. BM3011L
The Tris Buffered Saline (TBS) is an isotonic non-toxic buffer solution used commonly in biochemical applications. Tris maintains a stable pH range typically between 7.4 to 8.0 while NaCl regulates osmotic balance via its elected concentration. Tris maintains a stable pH range typically between 7.4 to 8.0 while NaCl regulates osmotic balance via its elected concentration. This TBS Solution is formulated at a 10X concentration, and is useful for washing in Western Blotting.