Learn More
Seracare Life Sciences Inc 10X TRIS-GLYCINE-SDS BUFFER
Catalog No. 5067473
Encompass_Preferred
Tris-Glycine-SDS (TGS) is a running buffer for sodium dodecyl sulfate – polyacrylamide gel electrophoresis (SDS-PAGE) of proteins. pH=8.3. Conductivity controlled.
This item is not returnable. View return policy
Catalog No. 50-674-73 Supplier Seracare Life Sciences Inc Supplier No. 54600016
Tris-Glycine-SDS (TGS) is a running buffer for sodium dodecyl sulfate – polyacrylamide gel electrophoresis (SDS-PAGE) of proteins. pH=8.3. Conductivity controlled.