Learn More
Seracare Life Sciences Inc 10X TRIS-GLYCINE TRANS BUFFER
Supplier:  Seracare Life Sciences Inc 54600011
Encompass_Preferred
10X Tris-glycine Transfer Buffer is intended for Western blotting and gel electrophoresis. It is ideal for preparing a standard Western blot transfer buffer (Towbin) and for use as a gel electrophoresis buffer for native Tris-glycine gel without SDS. pH=8.3. Conductivity controlled.
Catalog No. 50-674-70