Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Diagnostics Tris-Glycine-SDS PAGE Buffer (10X), 4 L, ultra-pure, clear, colorless solution
SDP

Catalog No. NC2307036
Encompass

The National Diagnostics EC-870 Tris-Glycine-SDS PAGE Buffer (10X) is an ultra-pure, concentrated solution designed for use as a running buffer in SDS-PAGE of proteins. This buffer is prepared with 18 MegOhm water and 0.2 micron filtration, saving significant preparation time and improving assay reproducibility, making it a cost-effective choice for laboratory settings.

  • Concentration of 10X
  • Used for SDS-PAGE running buffer
  • Target analyte is proteins
  • Volume of 4 L
  • Formulation purity is ultra-pure
  • Water purity of 18 MegOhm
  • Filtration of 0.2 micron
  • Available in liquid concentrate form
  • Clear, colorless solution
  • Handling precautions for skin, eye, and respiratory irritation

Catalog No. NC2307036 Supplier National Diagnostics Supplier No. EC870
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The National Diagnostics EC-870 Tris-Glycine-SDS PAGE Buffer (10X) is an ultra-pure, concentrated solution designed for use as a running buffer in SDS-PAGE of proteins. This buffer is prepared with 18 MegOhm water and 0.2 micron filtration, saving significant preparation time and improving assay reproducibility, making it a cost-effective choice for laboratory settings.

  • Concentration of 10X
  • Used for SDS-PAGE running buffer
  • Target analyte is proteins
  • Volume of 4 L
  • Formulation purity is ultra-pure
  • Water purity of 18 MegOhm
  • Filtration of 0.2 micron
  • Available in liquid concentrate form
  • Clear, colorless solution
  • Handling precautions for skin, eye, and respiratory irritation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.