Learn More
Rockland Immunochemicals 10X TTBS pH 7.5 (1.0 M Tris HCl 1.5 M Sodium Chloride 0.1% (v/v) Tween-20) - MB-013
SDP
Catalog No. 501058977
Encompass_Preferred
Tween Tris buffered saline is suitable for multiple applications including biological diluent buffer for antibodies or other biologics. Visit our newly expanded web site at www.rockland-inc.com for methods using this and other buffers. This product is a 10X concentrated stock solution and should be diluted appropriately with distilled, deionized water or equivalent to its final working concentration. This buffer consists of 1.0 M Tris HCl, 1.5 M Sodium Chloride and 0.1% (v/v) Tween-20 at a pH of 7.5. Meticulously prepared using ultra pure reagents dissolved in highly polished pharmaceutical grade deionized water.
Catalog No. 50-105-8977 Supplier Rockland Immunochemicals Supplier No. MB013
Tween Tris buffered saline is suitable for multiple applications including biological diluent buffer for antibodies or other biologics. Visit our newly expanded web site at www.rockland-inc.com for methods using this and other buffers. This product is a 10X concentrated stock solution and should be diluted appropriately with distilled, deionized water or equivalent to its final working concentration. This buffer consists of 1.0 M Tris HCl, 1.5 M Sodium Chloride and 0.1% (v/v) Tween-20 at a pH of 7.5. Meticulously prepared using ultra pure reagents dissolved in highly polished pharmaceutical grade deionized water.