Learn More
Enzo Life Sciences 10x Ubiquitinylation Buffer (5 ml)
SDP
Catalog No. NC1275802
Encompass_Preferred
Assay Buffer from the Ubiquitinylation Kits. Packaged as 5 x 1 ml. Shipping: Shipped on Blue Ice. Long Term Storage: -80°C
Catalog No. NC1275802 Supplier Enzo Life Sciences Supplier No. BMLKW98850005
Assay Buffer from the Ubiquitinylation Kits. Packaged as 5 x 1 ml. Shipping: Shipped on Blue Ice. Long Term Storage: -80°C