Learn More
ECM Bioscience 10X Western Blot Lysis Buffer, 15mL
Catalog No. NC1671658
Encompass
1% SDS Lysis Buffer is great for fully solubilizing proteins in preparation for Western blotting! It is suitable for both organs and tissue culture cells. Nonionic detergents such as NP-40 or triton fail to solubilize many cellular proteins involved in cell signaling. This problem is particularly acute in brain where synaptic junctions are known to be insoluble in nonionic detergents. 1% SDS completely solubilizes membrane and other hard to solubilize proteins including synaptic junction proteins. Suitable for both organs and tissue culture cells.
Catalog No. NC1671658 Supplier ECM Bioscience Supplier No. 100LYS
1% SDS Lysis Buffer is great for fully solubilizing proteins in preparation for Western blotting! It is suitable for both organs and tissue culture cells. Nonionic detergents such as NP-40 or triton fail to solubilize many cellular proteins involved in cell signaling. This problem is particularly acute in brain where synaptic junctions are known to be insoluble in nonionic detergents. 1% SDS completely solubilizes membrane and other hard to solubilize proteins including synaptic junction proteins. Suitable for both organs and tissue culture cells.