Learn More
MSC PRO-SOURCE Pleated Air Filter, MERV 8, 10x10x1 in, 1 ea
Catalog No. 5000502659
Encompass
This pleated air filter features a MERV 8 rating and synthetic media for efficient filtration. It is designed with a beverage board frame and is suitable for various applications, providing high capacity air purification for any unit.
  • Wireless pleated design
  • MERV 8 rating for efficient air filtration
  • Synthetic media material
  • Beverage board frame for durability
  • Nominal size of 10 in high, 10 in wide, 1 in deep
  • Air flow of 347 CFM
  • Maximum velocity of 500 FPM
  • UL 900 standards compliant
  • High capacity
Catalog No. 50-005-02659 Supplier MSC Supplier No. 14281174
This pleated air filter features a MERV 8 rating and synthetic media for efficient filtration. It is designed with a beverage board frame and is suitable for various applications, providing high capacity air purification for any unit.
  • Wireless pleated design
  • MERV 8 rating for efficient air filtration
  • Synthetic media material
  • Beverage board frame for durability
  • Nominal size of 10 in high, 10 in wide, 1 in deep
  • Air flow of 347 CFM
  • Maximum velocity of 500 FPM
  • UL 900 standards compliant
  • High capacity