Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC Value Collection Wheel Chock, Type A, 10 in, 1 ea

Catalog No. 502241485
Encompass

This Type A rubber wheel chock is designed to provide reliable support for securing wheels. It is constructed from durable rubber, offering quality and value for various business needs.

  • Type A wheel chock
  • Made from rubber
  • Features 10 in depth
  • Features 254 mm overall length

This item is not returnable. View return policy
Catalog No. 50-224-1485 Supplier MSC Supplier No. 51380731
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This Type A rubber wheel chock is designed to provide reliable support for securing wheels. It is constructed from durable rubber, offering quality and value for various business needs.

  • Type A wheel chock
  • Made from rubber
  • Features 10 in depth
  • Features 254 mm overall length

This item is not returnable. View return policy
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.