Learn More
Elkay Plastics Company Bag Reclosable Clear with Zip Closure 2 mil 10 X 12 1,000 per Case
Catalog No. NC0420601
Encompass_Preferred
LK Clear Line Zipper Reclosable bags provide excellent protection, convenience and economy. The zipper type seal closes securely and is easy to open and close time after time. These bags are excellent for storing, displaying, protecting and shipping all types of product. Because they are made only with virgin resin, they meet FDA requirements for contact with food products. Bags under 15" wide are packed in cartons of 1,000 with inner packs of 100 bags each for added convenience and to help reduce waste.
Catalog No. NC0420601 Supplier Elkay Plastics Company Supplier No. F21012
LK Clear Line Zipper Reclosable bags provide excellent protection, convenience and economy. The zipper type seal closes securely and is easy to open and close time after time. These bags are excellent for storing, displaying, protecting and shipping all types of product. Because they are made only with virgin resin, they meet FDA requirements for contact with food products. Bags under 15" wide are packed in cartons of 1,000 with inner packs of 100 bags each for added convenience and to help reduce waste.