Learn More
Integrated DNA Technologies Inc 10 x 2 mL IDTE pH 8.0 (1X TE Solution)
Catalog No. NC0911666
Encompass
IDTE (10 mM Tris, 0.1 mM EDTA) is our recommended solution for resuspending and storing single-stranded DNA and RNA oligos. It offers the most stability for the longest duration when compared to oligos stored dry or in water. IDTE is available at pH 7.5 or pH 8.0.
Catalog No. NC0911666 Supplier Integrated DNA Technologies Inc Supplier No. 11010205
This product needs to be quoted. Please contact your Fisher Scientific Sales Representative for pricing.
IDTE (10 mM Tris, 0.1 mM EDTA) is our recommended solution for resuspending and storing single-stranded DNA and RNA oligos. It offers the most stability for the longest duration when compared to oligos stored dry or in water. IDTE is available at pH 7.5 or pH 8.0.