Learn More
Ace Glass, Inc. Adjusta-chrom; column, 10mm ID x 300mm, complete with top& bottom adjusting plungers with PTFE seals and a 3 meter length of 1.5mm ID PTFE tubing
SDP
Catalog No. 50972142
Encompass_Preferred
10mm I.D. x 300mm Adjusta-Chrom chromatographic column used for recycling, ascending, or descending applications. Supplied with top & bottom adjusting plungers with PTFE seals, 25-50micron glass disc, and a 3m length of 1.5mm ID PTFE tubing.
Catalog No. 50-972-142 Supplier Ace Glass, Inc. Supplier No. 581503
10mm I.D. x 300mm Adjusta-Chrom chromatographic column used for recycling, ascending, or descending applications. Supplied with top & bottom adjusting plungers with PTFE seals, 25-50micron glass disc, and a 3m length of 1.5mm ID PTFE tubing.