Learn More
Ace Glass, Inc. Adjusta-Chrom Michel-Miller column, 10mm ID x 300mm, complete with adjustable, plunging top fitting, bottom fitting with1/4-28 tap and a 3 meter length of 1.5mm ID PTFE tubing
SDP
Catalog No. 50972155
Encompass_Preferred
Catalog No. 50-972-155 Supplier Ace Glass, Inc. Supplier No. 581634