Learn More
Ace Glass, Inc. Adjusta-Chrom; column, 10mm ID x 300mm, glass column only
SDP
Supplier:  Ace Glass, Inc. 581526
Encompass_Preferred
10mm I.D. x 300mm Adjusta-Chrom chromatographic column used for recycling, ascending or descending applications.
Catalog No. 50-972-147