Learn More
Charles River Laboratories 10X75MM FLINT GLS DEPYRO
Catalog No. NC9254362
Encompass
ENDOTOXIN-FREE TEST TUBES 50/pack Depyrogenated flint glass tubes wrapped in aluminum foil 10 x 75mm
Catalog No. NC9254362 Supplier Charles River Laboratories Supplier No. T200
ENDOTOXIN-FREE TEST TUBES 50/pack Depyrogenated flint glass tubes wrapped in aluminum foil 10 x 75mm