Learn More
Andwin Scientific 10X8X24 1.5ML SMARTBAG 1000CS
SDP
Catalog No. NC0132178
Encompass_Preferred
The poly bags by International features durability with its tubular construction, bottom seal, and thickness of 1.5 mil. The bags are made with of 100% virgin Low Density Polyethylene (LDPE) resin and are measured 25.4 x 20.3 x 61 cm or 10 x 8 x 24 in
Catalog No. NC0132178 Supplier Andwin Scientific Supplier No. 89070620
The poly bags by International features durability with its tubular construction, bottom seal, and thickness of 1.5 mil. The bags are made with of 100% virgin Low Density Polyethylene (LDPE) resin and are measured 25.4 x 20.3 x 61 cm or 10 x 8 x 24 in