Learn More
Ace Glass, Inc. Adjusta-Chrom; column, 10mm ID x 900mm, complete with top& bottom adjusting plungers with PTFE seals and a 3 meter length of 1.5mm ID PTFE tubing
SDP
Catalog No. 50972143
Encompass_Preferred
10mm I.D. x 900mm Adjusta-Chrom chromatographic column used for recycling, ascending or descending applications. Supplied with top & bottom adjusting plungers with PTFE seals, 25-50mic glass disc, and a 3m length of 1.5mm I.D. PTFE tubing.
Catalog No. 50-972-143 Supplier Ace Glass, Inc. Supplier No. 581507
10mm I.D. x 900mm Adjusta-Chrom chromatographic column used for recycling, ascending or descending applications. Supplied with top & bottom adjusting plungers with PTFE seals, 25-50mic glass disc, and a 3m length of 1.5mm I.D. PTFE tubing.