Learn More
Ace Glass, Inc. Adjusta-Chrom; column, 10mm ID x 900mm, glass column only
SDP
Catalog No. 50972148
Encompass_Preferred
10mm I.D. x 900mm Adjusta-Chrom chromatographic column used for recycling, ascending or descending applications.
Catalog No. 50-972-148 Supplier Ace Glass, Inc. Supplier No. 581530
10mm I.D. x 900mm Adjusta-Chrom chromatographic column used for recycling, ascending or descending applications.