Learn More
Chemglass Life Sciences Graded Seal, Quartz to Borosilicate, 10mm ID X 12mm OD
SDP
Catalog No. CGQ031009
Encompass_Preferred
Graded Seal, Quartz to Borosilicate, 10mm ID X 12mm OD. Graded seals have 3 inches of quartz sealed to 4 inches of borosilicate.
Catalog No. CGQ031009 Supplier Chemglass Life Sciences Supplier No. CGQ031009
Graded Seal, Quartz to Borosilicate, 10mm ID X 12mm OD. Graded seals have 3 inches of quartz sealed to 4 inches of borosilicate.